Learn More
Roboz Surgical NO5 DUMONT STAINLESS STEEL
SDP
Supplier:  Roboz Surgical RS5001
Encompass
Dumont Tweezers; Pattern #5; Tips Closing Flat On 4mm; .02 X .06mm Tip Size; Half Curve; Stainless Steel; 110mm Length; Material: Inox; Biologie Tips
Catalog No. NC9682664