Learn More
Medchemexpress LLC Noba | 55894-52-5 | 98.61% | 309.31 | 250 MG

Supplier: Medchemexpress LLC 250MGHY137799
NOBA is a synthetic chromogenic substrate used to detect the enzyme activity of AplTX-II and can be used in the research of phospholipid hydrolysis. It is for research use only.
- Solid appearance, white to off-white color
- Powder storage at -20°C for 3 years or 4°C for 2 years
- In-solvent storage at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO (100 mg/mL) with ultrasonic assistance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.