Learn More
Cayman Chemical Nobiletin, 478-01-3, 25 mg
SDP
Supplier:  Cayman Chemical 1542125
Encompass_Preferred
A natural polymethoxylated flavonoid and potent antioxidant with good bioavailability; has anti-inflammatory effects that are relevant to a host of diseases, including cancer, arthritis, and neurodegeneration; inhibits P-glycoprotein and BCRP (IC50 = 11.5 and 4.9 μM, respectively)
Catalog No. NC1062970