Learn More
Cayman Chemical Cayman Chemical |Nobiletin 2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one |50 mg | 15421-50 | Purity: ≥98% | 478-01-3 | FW: 402.4
SDP
Supplier:  Cayman Chemical 1542150
Encompass_Preferred
A natural polymethoxylated flavonoid and potent antioxidant with good bioavailability; has anti-inflammatory effects that are relevant to a host of diseases, including cancer, arthritis, and neurodegeneration; inhibits P-glycoprotein and BCRP (IC50 = 11.5 and 4.9 μM, respectively)
Catalog No. NC2872646