Learn More
Enzo Life Sciences NOC-7 (10mg). CAS: 146724-84-7
SDP
Supplier:  Enzo Life Sciences ALX430018M010
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of 0.1mM NOC-7 is 5 min. at 37°C in 0.1M PBS, pH 7.4. Alternative name: 1-Hydroxy-2-oxo-3-(N-3-methyl-aminopropyl)-3-methyl-1-triazene. Purity: ≥96% (HPLC). Appearance: White to off-white crystalline solid. Long Term Storage: -20°C.
Catalog No. 50-200-9150