Learn More
Abcam Nocardamine (Deferrio x amine E), Antibiotic agent, 5MG
Supplier:  Abcam AB1448425MG
Encompass_Preferred
MW 600.7 Da, Purity >95%. Antibiotic agent. Siderophore iron chelator. Induces morphological changes in insect cells. Shows weak antimicrobial against Gram-positive bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5483