Learn More
Medchemexpress LLC Nociceptin (Orphanin FQ) | 170713-75-4 | 98.7% | 1809.04 | 10 MG

Supplier: Medchemexpress LLC HYP018310MG
Nociceptin, a heptadecapeptide, is the endogenous ligand of the nociceptin receptor, acting as a potent anti-analgesic.
- Target: Opioid receptor
- Pathway: GPCR/G protein; neuronal signaling
- Sequence: Phe-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-Leu-Ala-Asn-Gln
- Appearance: Solid, white to off-white
- Shipping: Room temperature in continental US (may vary elsewhere)
- Storage (powder): -80°C for 2 years; -20°C for 1 year (sealed, away from moisture)
- Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.