Learn More
AdipoGen Nocodazole,Chemical. CAS: 31430-18-9
SDP
Supplier:  AdipoGen AGCR10019M025
Encompass_Preferred
Nocodazole, AdipoGen Life Sciences. Chemical. CAS: 31430-18-9. Formula: C14H11N3O3S. MW: 301.3. Microtubule inhibitor. Antitumor compound. Mitosis inhibitor. Arrests the cell cycle at G2/M phase. Promotes tubulin depolymerization. Induces fragmentation of the Golgi complex. Inhibits the T cell antigen receptor. Stimulates the intrinsic GTPase activity of tubulin. Activates the JNK/SAPK signaling pathway. Apoptosis inducer. Autophagy inhibition through prevention of autophagosome-lysosome fusion.
Catalog No. 50-464-566