Learn More
TARGETMOL CHEMICALS INC Nocodazole 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T280225MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Nocodazole (Oncodazole) is a reversible inhibitor of microtubule polymerization and an inhibitor of Bcr-Abl. Nocodazole has antitumor activity, blocks the cell cycle and induces apoptosis. Purity 99.34%
Catalog No. 50-287-8673