Learn More
Cayman Chemical NoctadecnoylLHomoserIn l 10mg
SDP
Supplier:  Cayman Chemical 1320910 MG
Encompass_Preferred
One of four lipophilic, long acyl side-chain bearing AHLs produced by the LuxI AHL synthase homolog SinI involved in quorum sensing signaling in strains of S. meliloti, a nitrogen-fixing bacterial symbiont of the legume M. sativa
Catalog No. 50-295-3032