Learn More
Medchemexpress LLC NODAGA-LM3 | 1415238-78-6 | 99.7% | 1521.11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP53621MG
Encompass_Preferred
NODAGA-LM3 can be labeled by 68Ga for PET imaging and is a SSTR2 antagonist. It can be used for imaging of SSTR positive paragangliomas. NODAGA-LM3 can also be labeled with [68Ga] for the synthesis/research of Radionuclide-Drug Conjugates (RDCs).
  • Target is SSTR2
  • Used in clinical trials for neuroendocrine tumors
  • Suitable for research use only
Catalog No. 50-003-96921