Learn More
Selleck Chemical LLC Nodakenin S9260-1MG
SDP
Supplier:  Selleck Chemical LLC S92601MG
Encompass_Preferred
Nodakenin (( )-Marmesinin) a coumarin compound acts as an AChE inhibitor that inhibits AChE activity in a dosedependent manner with an IC50 value of 84 7 M
Catalog No. 50-000-23785