Learn More
Hach Company NoElectrod Conduct Sens-San-PP
Supplier:  Hach Company 5000TC005000
Encompass
Electrodeless Conductivity Sensor-Sanitary Style-PP Body MaterialPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20ft) Analog Cable 3705E2T Electrodeless Conductivity Sensor
Catalog No. 50-002-07993