Learn More
Cayman Chemical noerIocItrIn 10mg
SDP
Supplier:  Cayman Chemical 2583910 MG
Encompass_Preferred
A flavonoid; inhibits the production of DPPH radicals by 17.2% and the formation of superoxide radicals by 48.3% in cell-free assays; decreases the rate of LDL oxidation ex vivo in isolated hamster plasma; increases proliferation of MC3T3-E1 cells and reverses the antiproliferative effect of PD 98059; increases the expression of the osteogenic differentiation markers Runx2, Type 1 collagen, and osteocalcin
Catalog No. 50-293-5527