Learn More
MSC Noga Mini PH Holder, Fine Adjustment, Magnetic Base, Universal Swivel Clamp
Supplier:  MSC 20173605
Encompass
The Mini PH Holder features an improved single-direction fine adjustment at its base. It includes a powerful on/off magnetic base and a universal swivel clamp designed for all dial and test indicators. The device ensures a strong arm lock using a unique 3-point system and is easy to set up for positioning gauges.
  • Improved single direction fine adjustment at base.
  • Powerful on/off magnetic base.
  • Universal swivel clamp for all dial and test indicators.
  • Strong arm lock by using the unique 3-point system.
  • Easy to set up gauges into required position.
Catalog No. 50-005-29526