Learn More
Biomatik Corporation Noggin (Nog) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29413
Encompass_Preferred
Noggin (Nog) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Noggin (NOG). The Noggin (Nog) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gln28~Cys232. Applications: WB, IHC. Target Synonyms: Noggin (NOG). Accession Number: Q13253. Gene ID: 9241. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4832