Learn More
Cayman Chemical NOLosartan A Other Receptor Mo
SDP
Supplier:  Cayman Chemical 100064561 MG
Encompass_Preferred
A potent AT1 receptor antagonist linked to an NO-donor; possesses similar anti-hypertensive effects to losartan, with the addition of the vasodilating effects of NO release
Catalog No. 50-288-9715