Learn More
Medchemexpress LLC HY-105634A 10mg Medchemexpress, Nomegestrol (acetate) CAS:58652-20-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105634A10MG
Encompass_Preferred
Medchemexpress, HY-105634A 10mg Nomegestrol (acetate) CAS:58652-20-3 Nomegestrol acetate is a potent, highly selective progestogen, which is characterized as a full agonist at the progesterone receptor, with no or minimal binding to other steroid receptors, including the androgen and glucocorticoid receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8283