Learn More
TARGETMOL CHEMICALS INC NOMIFENSINE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7308100MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nomifensine ((±)-Nomifensin) is a norepinephrine-dopamine reuptake inhibitor that prevents dopamine reuptake into synaptosomes. purity: 98%
Catalog No. 50-257-9223