Learn More
Medchemexpress LLC Nomilin | 1063-77-0 | 514.56 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN054710MM/1ML
Encompass_Preferred
Nomilin is a limonoid compound derived from citrus fruits, primarily functioning as an anti-obesity and anti-hyperglycemic agent.
  • Derived from citrus fruits
  • Acts as an anti-obesity agent
  • Functions as an anti-hyperglycemic agent
  • Mitigates cell death and reduces ROS production in SH-SY5Y cells
  • Attenuates blood-brain barrier disruption in MCAO rats
Catalog No. 50-003-88425