Learn More
Cayman Chemical nomycInsulfate 100g
SDP
Supplier:  Cayman Chemical 14287100 G
Encompass_Preferred
An aminoglycoside antibiotic that inhibits protein translation by binding to the small subunit of prokaryotic ribosomes; commonly used for the prevention of bacterial contamination of cell cultures
Catalog No. 50-295-5860