Learn More
CPC Scientific H-Glu-Gln-Val-Thr-Asn-Val-Gly-Gly-Ala-Val-Val-Thr-Gly-Val-Thr-Ala-Val-Ala-Gln-Lys-Thr-Val-Glu-Gly-Ala-Gly-Ser-Ile-Ala-Ala-Ala-Thr-Gly-Phe-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD052B
Encompass
This peptide was isolated from the insoluble core of Alzheimer's disease (AD) amyloid plaque. It is the fragment (61-95) of NACP or alpha-synuclein and forms amyloid fibrils via a nucleation-dependent polymerization mechanism. Accumulation of NAC aggregates in the synapse might be responsible for the neurodegeneration in AD and in the prion diseases.ONE-LETTER SEQUENCE: EQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVMOLECULAR FORMULA: C141H235N39O49MOLECULAR WEIGHT:3260.65STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [154040-19-4], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: H.Han et al., Chemistry and Biology, 2, 163 (1995); A.Iwai et al., Biochemistry, 34, 10139 (1995
Catalog No. 50-210-8715