Learn More
Medchemexpress LLC Nonadecane (Standard) | 629-92-5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN7928R
Encompass_Preferred
Nonadecane (Standard) is an analytical standard, 99.60% pure, intended for research and analytical applications. This reference standard is widely utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS. Nonadecane is sourced from plants, specifically Cactaceae Opuntia littoralis (Engelm.) Britton & Rose, and has also been identified for its use as a herbicide to control weeds in Dendrobium orchids.
  • Analytical standard for research and analytical applications.
  • High purity: 99.60%.
  • Versatile for use in HPLC, GC, and MS experiments.
  • Naturally derived from plants.
  • Acts as a herbicide for weed control in Dendrobium orchids.
Catalog No. 50-004-20570