Learn More
Medchemexpress LLC Nonadecanedioic acid | 6250-70-0 | MFCD00039536 | >98.0% | 328.5 g/mol | C19H36O4 | 1 G

Supplier: Medchemexpress LLC HYW0345961G
Nonadecanedioic acid (C19H36O4) is a long-chain saturated dicarboxylic acid used as a biochemical reagent and synthetic building block for research and analytical applications. It is typically handled as a white, waxy solid and is employed in organic synthesis and polymer chemistry as a precursor or standard.
- Long-chain saturated dicarboxylic acid (C19H36O4).
- Suitable as a building block for polymer and ester synthesis.
- Used as a reagent or standard in life-science and organic chemistry research.
- Typically supplied as a white waxy solid.
- Common purity greater than 98.0% as supplied by chemical vendors.
- Molecular weight approximately 328.5 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.