Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonaethylene glycol 5g | 3386-18-3 | 414.49 g·mol⁻¹ | C18H38O10 | 5 G
SDP

Supplier:  Medchemexpress LLC HY1168995G

Encompass_Preferred

Nonaethylene glycol is a linear polyethylene glycol (PEG) linker composed of nine ethylene glycol units. It is used as a flexible, hydrophilic spacer in PROTAC synthesis and other small-molecule conjugation chemistries, and is supplied for research use under recommended storage conditions.

  • Cas number 3386-18-3.
  • Molecular formula C18H38O10.
  • Molecular weight 414.49 g·mol⁻¹.
  • Typical use as a PEG-based PROTAC linker and spacer.
  • Available packaging 5 G, 10 G, 25 G.
  • Storage recommendations: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-27285


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.