Learn More
Medchemexpress LLC HY-N2334A 100mg Medchemexpress, Glycochenodeoxycholic acid (sodium salt) CAS:16564-43-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2334A100MG
Encompass_Preferred
Medchemexpress, HY-108294 100mg Nonaethylene glycol monododecyl ether CAS:3055-99-0 Nonaethylene glycol monododecyl ether (Nonaoxyethylene monododecyl ether) is a nonionic surfactant and polyethylene glycol (PEG) detergent that can be used to form initial coalesced O/W emulsion droplets, as well as for protein separation and purification[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0383