Learn More
Sigma Aldrich Fine Chemicals Biosciences Nonafluorobtyl methyl eth250ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 65139250ML
Encompass
consists of two inseparable isomers (Cf3)2CfCf2OCH3 and Cf3(Cf2)3OCH3 with almost identical properties
Catalog No. 50-289-7722