Learn More
Medchemexpress LLC Nonan-1-ol-d19 | 349553-86-2 | 98.0% | 163.37 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008816S10MG
Encompass_Preferred
n-Nonyl Alcohol-d19 is the deuterium labeled Nonan-1-ol. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53718