Learn More
Medchemexpress LLC Nonanoic acid-d2 | 62689-94-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN7057S350MG
Encompass_Preferred
Nonanoic acid-d2 is a deuterium labeled nonanoic acid, which is a naturally-occurring saturated fatty acid with nine carbon atoms. It has been shown to significantly reduce bacterial translocation, enhance antibacterial activity, and remarkably increase the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-18989