Learn More
Medchemexpress LLC Nonanoic acid-d3 | 134646-27-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN7057S15MG
Encompass_Preferred
Nonanoic acid-d3 is the deuterium-labeled form of nonanoic acid, a naturally occurring saturated fatty acid with nine carbon atoms. It has been shown to reduce bacterial translocation, enhance antibacterial activity, and increase the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Deuterium-labeled form of nonanoic acid
  • Naturally occurring saturated fatty acid
  • Reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 (pBD-1) and pBD-2
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-18689