Learn More
Medchemexpress LLC Nonapeptide-1 | 158563-45-2 | 98.74% | 1206.50 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP00975MG
Encompass_Preferred
Nonapeptide-1, also known as Melanostatine-5, is a peptide hormone acting as a selective antagonist of MC1R (Ki: 40 nM). It functions as a competitive α-MSH antagonist, effectively inhibiting intracellular cAMP and melanosome dispersion in melanocytes induced by α-MSH (IC50: 2.5 nM and 11 nM, respectively). This compound is valuable in research concerning skin pigmentation, regulation of steroid production in the adrenal gland, and skin cancer studies due to its ability to inhibit melanin synthesis.
  • Selective MC1R antagonist
  • Potently inhibits intracellular cAMP and melanosome dispersion
  • Inhibits melanin synthesis
  • Useful for skin pigmentation research
  • Applicable in studies of adrenal gland steroid regulation
  • Relevant to skin cancer research
Catalog No. 50-004-25893