Learn More
Abcam Nonivamide, TRPV1 agonist, 1G
Supplier:  Abcam AB1446521G
Encompass_Preferred
MW 293.4 Da, Purity >98%. TRPV1 agonist. Less potent Capsaicin (ab141000) analog. Induces Ca²+ release from ER. Blood-brain barrier permeable. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5528