Learn More
Cayman Chemical Nonpolar LIpId MIxture 1pl 1mL
SDP
Supplier:  Cayman Chemical 293761 ML
Encompass_Preferred
A mixture of non-polar lipids; contains cholesteryl palmitate, 1,2,3-tripalmitoyl-rac-glycerol, palmitic acid, and cholesterol; intended for use as a reference standard for the detection of these non-polar lipids in various sample types by TLC, LC-, or GC-MS
Catalog No. 50-299-0195