Learn More
SCANTIBODIES LABORATORY INC NON-SPECIFIC ANTIBODY BLOCKING TUBE (31X761) 100PK
Supplier:  SCANTIBODIES LABORATORY INC 31X761PK
Encompass
The 31X761 NABT (Non-Specific Antibody Blocking Tube) from Scantibodies Laboratory, Inc. contains immunoglobulins which the non-specific antibodies in the serum or plasma samples bind to, thus causing them to be blocked from interfering in antibody detection immunoassays. The NABT also allows for the rapid and simple elimination of false positive non-specific antibody interference in plasma or serum for detection assays (i.e., anti-HCV, HIV, Toxoplasmosis, Rubella, CMV, Herpes, Tg, Thyroglobulin, TPO Thrombopoietin, etc.). The reagent is in the form of a lyophilized pellet at the bottom of the tube and each tube contains enough reagents to inactivate the non-specific antibodies in 500 μl of sample material.
Catalog No. NC0393833