Learn More
Sigma Aldrich Fine Chemicals Biosciences Nonyl beta D glucopyranosi1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N75071G
Encompass
Nonyl -D-glucopyranoside is a nonionic surfactant among a small number of surfactants found to be useful for the solubilization and crystallization of a large number of biological membrane proteins.
Catalog No. 50-290-2428