Learn More
TARGETMOL CHEMICALS INC Nootkatone 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T3S01951G
Encompass_Preferred
Also available in 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Nootkatone can provide effective tick control. 2. (+)-Nootkatone has antiallergic, anti-inflammatory, and antiplatelet activities. 3. Nootkatone can prevent obesity and improve physical performance through AMPK activation in skeletal muscle and liver. Purity 99.04%
Catalog No. 50-287-6383