Learn More
ATCC NOR-10; Muscle; Mouse (Mus musculus)
Supplier:  ATCC CCL197
Encompass
NOR-10 cells are fibroblasts derived from the abdominal wall muscle of a normal mouse. This cell line can be used as a matched control for BLO-11 (ATCC CCL-198).
Catalog No. 50-238-2092