Learn More
Apexbio Technology LLC Noradrenaline bitartrate monohydrate 108341-18-0 500mg
SDP
Supplier:  Apexbio Technology LLC B1990500
Encompass_Preferred
Noradrenaline bitartrate monohydrate (CAS 108341-18-0) is an adrenergic receptor agonist targeting - - and -adrenoceptors It is designed to activate these receptors thereby modulating adrenergic signaling pathways Noradrenaline bitartrate monohydrate exerts its biological activity primarily through -adrenoceptor-mediated vasoconstriction in peripheral vasculature It demonstrates receptor subtype selectivity with Ki values of approximately 330 nM ( ) 56 nM ( ) and 740 nM ( ) Based on these pharmacological properties noradrenaline bitartrate monohydrate holds research potential in elucidating adrenergic receptor-mediated physiological and pathophysiological mechanisms in neurological and cardiovascular contexts
Catalog No. 50-000-92874