Learn More
Medchemexpress LLC Norartocarpetin | 520-30-9 | 286.24 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN105031MG
Encompass_Preferred
Norartocarpetin is a potent tyrosinase inhibitor (IC50 = 0.47 μM) and can act as an antibrowning agent in food research. It shows significant anticancer activity in lung carcinoma cells (NCI-H460, IC50 = 22 μM), mediating antiproliferative effects via the Ras/Raf/MAPK pathway, mitochondrial apoptosis, S-phase cell cycle arrest, and suppression of cell migration/invasion.
  • Tyrosinase inhibitor (IC50 = 0.47 μM).
  • Antibrowning agent for food systems research.
  • Anticancer activity in lung carcinoma cells (NCI-H460, IC50 = 22 μM).
  • Antiproliferative effects via Ras/Raf/MAPK pathway.
  • Induces mitochondrial-mediated apoptosis.
  • Causes S-phase cell cycle arrest.
  • Suppresses cell migration and invasion.
Catalog No. 50-004-15601