Learn More
Cayman Chemical NorcholIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3462310 MG
Encompass_Preferred
A bile acid and derivative of cholic acid; urine levels are increased in patients with liver cirrhosis or CTX
Catalog No. 50-294-8042