Learn More
Medchemexpress LLC Norcholic acid | 60696-62-0 | 98.8% | 394.54 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN94575MG
Encompass_Preferred
Norcholic acid is a normal minorbile C23 bile acid with four side chains, naturally occurring in human urine and meconium. Its presence and levels can become more prominent under certain pathological conditions. This compound is known for being efficiently absorbed from the intestine and subsequently rapidly excreted into the bile, although it does not typically excrete into urine.
  • Normal minorbile C23 bile acid
  • Found in human urine and meconium
  • Levels may increase in pathological conditions
  • Efficiently absorbed from the intestine
  • Rapidly excreted into the bile
Catalog No. 50-004-26094