Learn More
Medchemexpress LLC Norcholic acid | 60696-62-0 | 99.15% | 394.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN945710MM/1ML
Encompass_Preferred
Norcholic acid is a normal minorbile C23 bile acid with four side chains, found in human urine and meconium. It can become prominent under certain pathological conditions. This compound is efficiently absorbed from the intestine and quickly excreted into the bile, but not into urine.
  • Normal minorbile C23 bile acid
  • Four side chains
  • Found in human urine and meconium
  • Can become prominent under certain pathological conditions
  • Efficiently absorbed from the intestine
  • Quickly excreted into the bile
  • Not excreted into urine
  • In vivo studies show approximately 40% of administered radioactivity recovered from feces within 24 hours in rats
Catalog No. 50-004-26480