Learn More
Medchemexpress LLC Norcholic acid | 60696-62-0 | 99.2% | 394.54 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN945710MG
Encompass_Preferred
Norcholic acid is a normal minor C23 bile acid that possesses a four-side chain structure and can be found in human urine and meconium. While typically a minor component, its presence can become notable under certain pathological conditions. This compound is effectively absorbed from the intestine and promptly excreted into the bile, but not into the urine.
  • It is a normal minor C23 bile acid.
  • It has a four side chain structure.
  • It exists in human urine and meconium.
  • It can become prominent under certain pathological conditions.
  • It is efficiently absorbed from the intestine.
  • It is quickly excreted into the bile.
  • It is not excreted into urine.
Catalog No. 50-004-26345