Learn More
Medchemexpress LLC Norcyclizine (1-(diphenylmethyl)piperazine) | 841-77-0 | 99.7% | 252.35 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013376100G
Encompass_Preferred
Norcyclizine is a piperazine compound used for the synthesis of antimicrobial agents. 1-Benzhydrylpiperazine derivatives, including Norcyclizine, have been shown to enhance the antibacterial activity of β-lactam antibiotics against Methicillin-resistant Staphylococcus aureus (MRSA). This enhancement is believed to occur through the inhibition of the allosteric site of PBP2a. Additionally, 1-Benzhydrylpiperazine can function as a pharmacological scaffold for the synthesis of anticancer agents.
  • Can be used for the synthesis of antimicrobial agents.
  • Enhances the antibacterial activity of β-lactam antibiotics against Methicillin-resistant Staphylococcus aureus (MRSA) by inhibiting the allosteric site of PBP2a.
  • Can serve as a pharmacological scaffold for the synthesis of anticancer agents.
Catalog No. 50-004-54624