Learn More
Medchemexpress LLC Nordeoxycholic acid | 53608-86-9 | 10 MG

Supplier: Medchemexpress LLC HYN826810MG
Nordeoxycholic acid is a 23-carbon bile acid. It is a norcholic acid metabolite and a steroid human metabolite.
- Purity: 99.90%
- Molecular weight: 378.55
- Formula: C23H38O4
- Appearance: Solid
- Color: White to off-white
- Initial source: Animals, bile
- Storage: -20°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
- Solubility: DMSO: 100 mg/mL (264.17 mM, requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.