Learn More
Medchemexpress LLC Nordeoxycholic acid | 53608-86-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN82685MG
Encompass_Preferred
Nordeoxycholic acid is a 23-carbon bile acid, a norcholic acid metabolite, and a steroid human metabolite, intended for research use only.
  • A 23-carbon bile acid
  • Norcholic acid metabolite
  • Steroid human metabolite
  • High purity (99.90%)
  • White to off-white solid
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-19223