Learn More
Selleck Chemical LLC Nordihydroguaiaretic acid (NDGA) S3984-25mg
SDP
Supplier:  Selleck Chemical LLC S398425MG
Encompass_Preferred
Nordihydroguaiaretic acid (NDGA) is a phenolic antioxidant found in the leaves and twigs of the evergreen desert shrub Larrea tridentata (Sesse and Moc ex DC) Coville (creosote bush) It is a recognized inhibitor of lipoxygenase (LOX) and has antioxidant and free radical scavenging properties Nordihydroguaiaretic acid (NDGA) is a cytotoxic insulin-like growth factor-I receptor (IGF-1R)/HER2 inhibitor and induces apoptosis
Catalog No. 50-310-8152