Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | MFCD00135797 | 99.9% | 301.81 | C18H20ClNO | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1000461MG

Encompass_Preferred

Nordoxepin hydrochloride is the major metabolite of the tricyclic antidepressant doxepin. Supplied as a high-purity chemical standard for laboratory research and analytical applications, this compound is intended for research use only.

  • Major metabolite of doxepin, useful in metabolism and analytical studies.
  • Purity 99.94% suitable for analytical applications.
  • Solid, white to off-white; easy to handle and store.
  • Molecular formula C18H20ClNO and molecular weight 301.81 g/mol.
  • Storage: sealed, away from moisture; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in multiple package sizes and solution forms for experimental flexibility.

Catalog No. 50-002-15208


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.