Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | MFCD00135797 | 99.9% | 301.81 g·mol⁻¹ | C18H20ClNO | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10004610MM 1ML

Encompass_Preferred

Nordoxepin hydrochloride is the hydrochloride salt of nordoxepin (desmethyldoxepin), the primary metabolite of the tricyclic antidepressant doxepin. It is intended for research and analytical applications and is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid mass formats for analytical and synthetic workflows.

  • High purity suitable for research applications (99.94%).
  • Supplied as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Available in multiple solid mass sizes for synthetic and analytical work.
  • Molecular weight 301.81 g·mol⁻¹ and formula C18H20ClNO for easy reference.
  • Storage guidance: solid at 4°C; solution at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-15656


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.