Learn More
Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | MFCD00135797 | 99.9% | 301.81 g/mol | C18H20ClNO | 100 MG

Supplier: Medchemexpress LLC HY100046100MG
Nordoxepin hydrochloride is the hydrochloride salt of nordoxepin (desmethyldoxepin), the primary metabolite of the tricyclic antidepressant doxepin. Supplied as a high-purity solid reference standard for analytical and preclinical research, it is intended for metabolite identification, method development, and quantitative assay use. Not for human or clinical use.
- High purity (99.94%).
- CAS number 2887-91-4.
- Chemical formula C18H20ClNO.
- Molecular weight 301.81 g/mol.
- White to off-white solid appearance.
- Store sealed, away from moisture at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.